accessibility menu, dialog, popup

UserName

CAS RN 31127-80-7

IUPAC Name: N1,N3-bis(2,3-dihydroxypropyl)-5-acetamido-2,4,6-triiodobenzene-1,3-dicarboxamide
Synonyms: N1,N3-bis(2,3-dihydroxypropyl)-5-acetamido-2,4,6-triiodobenzene-1,3-dicarboxamide
Molecular Weight (g/mol): 747.06
Molecular Formula: C16H20I3N3O7
InChi Key: BHCBLTRDEYPMFZ-UHFFFAOYNA-N
SMILES: CC(=O)NC1=C(I)C(C(=O)NCC(O)CO)=C(I)C(C(=O)NCC(O)CO)=C1I
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results