accessibility menu, dialog, popup

UserName

CAS RN 307496-27-1

IUPAC Name: (2-bromophenyl)methanide; bromozincylium
Synonyms: (2-bromophenyl)methanide; bromozincylium
Molecular Weight (g/mol): 315.31
Molecular Formula: C7H6Br2Zn
InChi Key: DGWAMZSHJOIPLU-UHFFFAOYSA-M
SMILES: [Zn+]Br.[CH2-]C1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results