accessibility menu, dialog, popup

UserName

CAS RN 3073-66-3

CH3H3CCH3

CAS RN 3073-66-3

IUPAC Name: 1,1,3-trimethylcyclohexane
Synonyms: 1,1,3-trimethylcyclohexane
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: PYOLJOJPIPCRDP-UHFFFAOYNA-N
SMILES: CC1CCCC(C)(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results