accessibility menu, dialog, popup

UserName

CAS RN 3068-34-6

IUPAC Name: [3,4-bis(acetyloxy)-6-chloro-5-acetamidooxan-2-yl]methyl acetate
Synonyms: [3,4-bis(acetyloxy)-6-chloro-5-acetamidooxan-2-yl]methyl acetate
Molecular Weight (g/mol): 365.76
Molecular Formula: C14H20ClNO8
InChi Key: NAYYKQAWUWXLPD-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(Cl)OC(COC(C)=O)C(OC(C)=O)C1OC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results