accessibility menu, dialog, popup

UserName

CAS RN 305-53-3

IUPAC Name: sodium 2-iodoacetate
Synonyms: sodium iodoacetate
Molecular Weight (g/mol): 207.93
Molecular Formula: C2H2INaO2
InChi Key: AGDSCTQQXMDDCV-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)CI

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results