accessibility menu, dialog, popup

UserName

CAS RN 303-25-3

IUPAC Name: hydrogen 1-(diphenylmethyl)-4-methylpiperazine chloride
Synonyms: hydrogen cyclizine chloride
Molecular Weight (g/mol): 302.85
Molecular Formula: C18H23ClN2
InChi Key: UKPBEPCQTDRZSE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results