accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHNNSOHO

CAS RN 302964-02-9

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-1,3-thiazole-5-carboxylic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 244.27
Molecular Formula: C9H12N2O4S
InChi Key: QNFLEDLPOVONCN-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=NC=C(S1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results