accessibility menu, dialog, popup

UserName

ONOOOO

CAS RN 3027-38-1

IUPAC Name: 11-nitro-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Synonyms: 11-nitro-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Molecular Weight (g/mol): 243.17
Molecular Formula: C12H5NO5
InChi Key: FLFLZYYDLIKGJQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=C3C(C=CC=C3C(=O)OC2=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results