accessibility menu, dialog, popup

UserName

BOOCH3H3CCH3CH3CH3CH3

CAS RN 302554-81-0

IUPAC Name: 2-(9,9-dioctyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(9,9-dioctylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 516.62
Molecular Formula: C35H53BO2
InChi Key: VOASJDUTCQKQLE-UHFFFAOYSA-N
SMILES: CCCCCCCCC1(CCCCCCCC)C2=CC=CC=C2C2=C1C=C(C=C2)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results