CAS RN 298-00-0
CAS RN 298-00-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl Parathion, SPEX CertiPrep™
CAS: 298-00-0 Molecular Formula: C8H10NO5PS Molecular Weight (g/mol): 263.204 InChI Key: RLBIQVVOMOPOHC-UHFFFAOYSA-N PubChem CID: 4130 ChEBI: CHEBI:38746 IUPAC Name: dimethoxy-(4-nitrophenoxy)-sulfanylidene-$l^{5}-phosphane SMILES: COP(=S)(OC)OC1=CC=C(C=C1)[N+](=O)[O-]