accessibility menu, dialog, popup

UserName

CAS RN 29681-98-9

CH3OHOF(Z)

CAS RN 29681-98-9

IUPAC Name: (3Z)-4-(4-fluorophenyl)-4-hydroxybut-3-en-2-one
Synonyms: (3Z)-4-(4-fluorophenyl)-4-hydroxybut-3-en-2-one
Molecular Weight (g/mol): 180.18
Molecular Formula: C10H9FO2
InChi Key: NVKHIXOPLVAELL-POHAHGRESA-N
SMILES: CC(=O)\C=C(/O)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results