accessibility menu, dialog, popup

UserName

CH3ONHSOONHOH3C

CAS RN 29591-86-4

IUPAC Name: N-(4-acetamidobenzenesulfonyl)acetamide
Synonyms: N-(4-acetamidobenzenesulfonyl)acetamide
Molecular Weight (g/mol): 256.28
Molecular Formula: C10H12N2O4S
InChi Key: YTJJPFRVBNFIBN-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results