accessibility menu, dialog, popup

UserName

H3COCH3NOONOO

CAS RN 29455-11-6

IUPAC Name: 2-methoxy-5-methyl-1,3-dinitrobenzene
Synonyms: 2-methoxy-5-methyl-1,3-dinitrobenzene
Molecular Weight (g/mol): 212.16
Molecular Formula: C8H8N2O5
InChi Key: HVNUPXQONRHUOX-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C)C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results