accessibility menu, dialog, popup

UserName

H3COH3CH3C

CAS RN 2944-49-2

IUPAC Name: 1-methoxy-2,3-dimethylbenzene
Synonyms: 2,3-dimethylanisole
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: BLMBNEVGYRXFNA-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C)=C1C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)