accessibility menu, dialog, popup

UserName

NH2SNH2NN

CAS RN 2937-81-7

IUPAC Name: 1,3,4-thiadiazole-2,5-diamine
Synonyms: 1,3,4-thiadiazole-2,5-diamine
Molecular Weight (g/mol): 116.14
Molecular Formula: C2H4N4S
InChi Key: DXVLLEIKCNQUQH-UHFFFAOYSA-N
SMILES: NC1=NN=C(N)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results