CAS RN 2929-52-4
IUPAC Name:
trihexylsilyl
Synonyms:
trihexylsilyl
Molecular Weight (g/mol):
283.6
Molecular Formula:
C18H39Si
InChi Key:
ISPSHPOFLYFIRR-UHFFFAOYSA-N
SMILES:
CCCCCC[Si](CCCCCC)CCCCCC
1
–
1
of
1
results
1
Trihexylsilane 95.0+%, TCI America™
CAS: 2929-52-4 Molecular Formula: C18H39Si Molecular Weight (g/mol): 283.595 MDL Number: MFCD00009519 InChI Key: ISPSHPOFLYFIRR-UHFFFAOYSA-N PubChem CID: 6327464 IUPAC Name: trihexylsilicon SMILES: CCCCCC[Si](CCCCCC)CCCCCC