CAS RN 29232-39-1
IUPAC Name:
2,3-dibromo-5-methylpyridine
Synonyms:
2,3-dibromo-5-methylpyridine
Molecular Weight (g/mol):
250.92
Molecular Formula:
C6H5Br2N
InChi Key:
BZUZAZMAGVFIBS-UHFFFAOYSA-N
SMILES:
CC1=CN=C(Br)C(Br)=C1