accessibility menu, dialog, popup

UserName

CAS RN 29089-62-1

BH3CPCH3H3CH3C

CAS RN 29089-62-1

IUPAC Name: tetrabutylphosphanium; tetraphenylboranuide
Synonyms: tetrabutylphosphonium tetraphenylborate
Molecular Weight (g/mol): 578.67
Molecular Formula: C40H56BP
InChi Key: OGWSZNMGVXKACZ-UHFFFAOYSA-N
SMILES: CCCC[P+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results