accessibility menu, dialog, popup

UserName

ON

CAS RN 28957-72-4

IUPAC Name: 8-benzyl-8-azabicyclo[3.2.1]octan-3-one
Synonyms: 8-benzyl-8-azabicyclo[3.2.1]octan-3-one
Molecular Weight (g/mol): 215.3
Molecular Formula: C14H17NO
InChi Key: RSUHKGOVXMXCND-UHFFFAOYNA-N
SMILES: O=C1CC2CCC(C1)N2CC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results