accessibility menu, dialog, popup

UserName

H3CBrCH3

CAS RN 287493-15-6

IUPAC Name: 2-bromo-9,9-diethyl-9H-fluorene
Synonyms: 2-bromo-9,9-diethylfluorene
Molecular Weight (g/mol): 301.23
Molecular Formula: C17H17Br
InChi Key: HJXPGCTYMKCLTR-UHFFFAOYSA-N
SMILES: CCC1(CC)C2=CC=CC=C2C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results