accessibility menu, dialog, popup

UserName

H3CN

CAS RN 28623-46-3

IUPAC Name: nonadecanenitrile
Synonyms: nonadecanenitrile
Molecular Weight (g/mol): 279.51
Molecular Formula: C19H37N
InChi Key: MVDGSHXQMOKTDJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCC#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results