accessibility menu, dialog, popup

UserName

CH3OSSOCH3

CAS RN 28588-75-2

IUPAC Name: 2-methyl-3-[(2-methylfuran-3-yl)disulfanyl]furan
Synonyms: 2-methyl-3-[(2-methylfuran-3-yl)disulfanyl]furan
Molecular Weight (g/mol): 226.31
Molecular Formula: C10H10O2S2
InChi Key: OHDFENKFSKIFBJ-UHFFFAOYSA-N
SMILES: CC1=C(SSC2=C(C)OC=C2)C=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results