accessibility menu, dialog, popup

UserName

CH3CH3HOH2N

CAS RN 2854-16-2

IUPAC Name: 1-amino-2-methylpropan-2-ol
Synonyms: 1-amino-2-methylpropan-2-ol
Molecular Weight (g/mol): 89.14
Molecular Formula: C4H11NO
InChi Key: LXQMHOKEXZETKB-UHFFFAOYSA-N
SMILES: CC(C)(O)CN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results