CAS RN 28226-22-4
IUPAC Name:
6-nitro-3,4-dihydro-2H-1,4-benzoxazine
Synonyms:
6-nitro-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol):
180.16
Molecular Formula:
C8H8N2O3
InChi Key:
GZAJZBARYACGSO-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C2OCCNC2=C1
1
–
1
of
1
results
1
Promotions Available
6-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%
6-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%
CAS: 28226-22-4 Molecular Formula: C8H8N2O3 Molecular Weight (g/mol): 180.163 MDL Number: MFCD07698591 InChI Key: GZAJZBARYACGSO-UHFFFAOYSA-N Synonym: 6-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine,2h-1,4-benzoxazine,3,4-dihydro-6-nitro,2h-1,4-benzoxazine, 3,4-dihydro-6-nitro,6-nitro-2,3-dihydrobenz-1,4-oxazine,6-nitro-2h,3h,4h-benzo e 1,4-oxazine,6-nitro-3,4-dihydro-2h-1,4 benzoxazine,6-nitro-3,4-dihydro-2h-benzo 1,4 oxazine,3,4-dihydro-6-nitro-2h-benzo b 1,4 oxazine,1-1,3 dioxolan-2-ylmethyl-1h-pyrazole-4-boronicacid,pinacolester PubChem CID: 7062225 IUPAC Name: 6-nitro-3,4-dihydro-2H-1,4-benzoxazine SMILES: C1COC2=C(N1)C=C(C=C2)[N+](=O)[O-]