accessibility menu, dialog, popup

UserName

H3CH3CCH3H3CCH3OHHO

CAS RN 28122-52-3

IUPAC Name: 4-(2,4,4-trimethylpentan-2-yl)benzene-1,3-diol
Synonyms: 4-(2,4,4-trimethylpentan-2-yl)benzene-1,3-diol
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: YQOJNENEFSZINP-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)C1=CC=C(O)C=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results