accessibility menu, dialog, popup

UserName

CH3SiOH3COH3COH3C

CAS RN 2761-24-2

IUPAC Name: triethoxy(pentyl)silane
Synonyms: n-pentyltriethoxysilane
Molecular Weight (g/mol): 234.41
Molecular Formula: C11H26O3Si
InChi Key: FHVAUDREWWXPRW-UHFFFAOYSA-N
SMILES: CCCCC[Si](OCC)(OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results