accessibility menu, dialog, popup

UserName

H3COHNSOOH

CAS RN 27486-87-9

IUPAC Name: 2-acetamido-3-[(triphenylmethyl)sulfanyl]propanoic acid
Synonyms: 2-acetamido-3-[(triphenylmethyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 405.51
Molecular Formula: C24H23NO3S
InChi Key: KCVPASSMLHHOIF-UHFFFAOYNA-N
SMILES: CC(=O)NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)