CAS RN 27326-43-8
IUPAC Name:
2-ethenylbenzoic acid
Synonyms:
2-ethenylbenzoic acid
Molecular Weight (g/mol):
148.16
Molecular Formula:
C9H8O2
InChi Key:
XUDBVJCTLZTSDC-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC=C1C=C
1
–
1
of
1
results
1
Promotions Available
2-Vinylbenzoic acid, 96%
2-Vinylbenzoic acid, 96%
CAS: 27326-43-8 Molecular Formula: C9H8O2 Molecular Weight (g/mol): 148.161 MDL Number: MFCD02066273 InChI Key: XUDBVJCTLZTSDC-UHFFFAOYSA-N PubChem CID: 3015504 IUPAC Name: 2-ethenylbenzoic acid SMILES: C=CC1=CC=CC=C1C(=O)O