accessibility menu, dialog, popup

UserName

CAS RN 2708-97-6

FFFIIF

CAS RN 2708-97-6

IUPAC Name: 1,2,3,4-tetrafluoro-5,6-diiodobenzene
Synonyms: 1,2,3,4-tetrafluoro-5,6-diiodobenzene
Molecular Weight (g/mol): 401.87
Molecular Formula: C6F4I2
InChi Key: JQBYIZAYQMMVTO-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(I)C(I)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)