accessibility menu, dialog, popup

UserName

H3CH2N

CAS RN 2696-85-7

IUPAC Name: 2-butylaniline
Synonyms: 2-butylaniline
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: HDVUPIFFKAHPJY-UHFFFAOYSA-N
SMILES: CCCCC1=CC=CC=C1N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results