CAS RN 268568-11-2
IUPAC Name:
ethyl 2-amino-5-iodobenzoate
Synonyms:
ethyl 2-amino-5-iodobenzoate
Molecular Weight (g/mol):
291.09
Molecular Formula:
C9H10INO2
InChi Key:
FPCLHSGOJPEKKE-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(N)C=CC(I)=C1
1
–
1
of
1
results
1
Promotions Available
Ethyl 2-amino-5-iodobenzoate, 98+%
Ethyl 2-amino-5-iodobenzoate, 98+%
CAS: 268568-11-2 Molecular Formula: C9H10INO2 Molecular Weight (g/mol): 291.09 MDL Number: MFCD04038940 InChI Key: FPCLHSGOJPEKKE-UHFFFAOYSA-N Synonym: ethyl2-amino-5-iodobenzoate,2-amino-5-iodobenzoic acid ethyl ester,2-amino-5-iodo-benzoic acid ethyl ester,acmc-1cksm,ethyl 2-amino-5-iodo-benzoate,ethyl 5-iodoanthranilate PubChem CID: 12012179 IUPAC Name: ethyl 2-amino-5-iodobenzoate SMILES: CCOC(=O)C1=C(N)C=CC(I)=C1