accessibility menu, dialog, popup

UserName

CAS RN 26549-22-4

CH3O(R)NHH3CCH3(R)NHCH3CH3OCH3

CAS RN 26549-22-4

IUPAC Name: [(2R,3R)-4-(dimethylazaniumyl)-2,3-dimethoxybutyl]dimethylazanium
Synonyms: [(2R,3R)-4-(dimethylammonio)-2,3-dimethoxybutyl]dimethylazanium
Molecular Weight (g/mol): 206.33
Molecular Formula: C10H26N2O2
InChi Key: VYQCQNCBTMHEFI-NXEZZACHSA-P
SMILES: CO[C@H](C[NH+](C)C)[C@@H](C[NH+](C)C)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results