accessibility menu, dialog, popup

UserName

ClClO

CAS RN 2648-61-5

IUPAC Name: 2,2-dichloro-1-phenylethan-1-one
Synonyms: 2,2-dichloro-1-phenylethanone
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: CERJZAHSUZVMCH-UHFFFAOYSA-N
SMILES: ClC(Cl)C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results