accessibility menu, dialog, popup

UserName

H3COClNN

CAS RN 26452-81-3

IUPAC Name: 4-chloro-6-methoxypyrimidine
Synonyms: 4-chloro-6-methoxypyrimidine
Molecular Weight (g/mol): 144.56
Molecular Formula: C5H5ClN2O
InChi Key: KLJGSQVYUGQOAW-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results