accessibility menu, dialog, popup

UserName

HON

CAS RN 2629-72-3

IUPAC Name: 3-(pyridin-4-yl)propan-1-ol
Synonyms: 4-pyridinepropanol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: PZVZGDBCMQBRMA-UHFFFAOYSA-N
SMILES: OCCCC1=CC=NC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)