accessibility menu, dialog, popup

UserName

CAS RN 2622-08-4

CH3OPOH3COH3C

CAS RN 2622-08-4

IUPAC Name: tris(2-methylphenyl) phosphite
Synonyms: tri-O-cresyl phosphite
Molecular Weight (g/mol): 352.37
Molecular Formula: C21H21O3P
InChi Key: BKHZQJRTFNFCTG-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OP(OC1=CC=CC=C1C)OC1=CC=CC=C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results