CAS RN 26218-80-4
IUPAC Name:
methyl 6-methoxypyridine-3-carboxylate
Synonyms:
methyl 6-methoxypyridine-3-carboxylate
Molecular Weight (g/mol):
167.16
Molecular Formula:
C8H9NO3
InChi Key:
OVLDWZNVBDRZNN-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(OC)N=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 6-methoxynicotinate, 98%
Methyl 6-methoxynicotinate, 98%
CAS: 26218-80-4 Molecular Formula: C8H9NO3 Molecular Weight (g/mol): 167.164 MDL Number: MFCD00674130 InChI Key: OVLDWZNVBDRZNN-UHFFFAOYSA-N Synonym: methyl 6-methoxynicotinate,6-methoxynicotinic acid methyl ester,methyl 2-methoxypyridine-5-carboxylate,3-pyridinecarboxylic acid, 6-methoxy-, methyl ester,methyl 2-methoxy-5-pyridinecarboxylate,2-methoxy-5-methoxycarbonyl pyridine,pubchem14334,acmc-1cdoo,methyl-6-methoxynicotinate,methyl6-methoxy-nicotinate PubChem CID: 586039 IUPAC Name: methyl 6-methoxypyridine-3-carboxylate SMILES: COC1=NC=C(C=C1)C(=O)OC