accessibility menu, dialog, popup

UserName

CAS RN 2617-79-0

IUPAC Name: N-(4-chlorophenyl)formamide
Synonyms: P-chloroformanilide
Molecular Weight (g/mol): 155.58
Molecular Formula: C7H6ClNO
InChi Key: LMLFHXMNNHGRRO-UHFFFAOYSA-N
SMILES: ClC1=CC=C(NC=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results