CAS RN 26163-03-1
CAS RN 26163-03-1
IUPAC Name:
3-bromo-5-chloropyridin-2-amine
Synonyms:
3-bromo-5-chloropyridin-2-amine
Molecular Weight (g/mol):
207.46
Molecular Formula:
C5H4BrClN2
InChi Key:
UWGGGYYCKDCTGN-UHFFFAOYSA-N
SMILES:
NC1=NC=C(Cl)C=C1Br
1
–
1
of
1
results
1
2-Amino-3-bromo-5-chloropyridine 98.0+%, TCI America™
CAS: 26163-03-1 Molecular Formula: C5H4BrClN2 Molecular Weight (g/mol): 207.455 MDL Number: MFCD00955627 InChI Key: UWGGGYYCKDCTGN-UHFFFAOYSA-N Synonym: 2-amino-3-bromo-5-chloropyridine,3-bromo-5-chloro-2-pyridinamine,2-pyridinamine, 3-bromo-5-chloro,3-bromo-5-chloro-2-pyridylamine,3-bromo-5-chloropyrid-2-ylamine,3-bromo-5-chloro-pyridin-2-ylamine,abbypharma ap-10-5406,3-bromo-5-chloro-pyridin-2-amine,pubchem1141,acmc-1ch3b PubChem CID: 2763971 IUPAC Name: 3-bromo-5-chloropyridin-2-amine SMILES: C1=C(C=NC(=C1Br)N)Cl