accessibility menu, dialog, popup

UserName

OSOOHO

CAS RN 260063-21-6

IUPAC Name: 2H,3H-thieno[3,4-b][1,4]dioxine-5-carboxylic acid
Synonyms: 2H,3H-thieno[3,4-b][1,4]dioxine-5-carboxylic acid
Molecular Weight (g/mol): 186.18
Molecular Formula: C7H6O4S
InChi Key: DCCRIIFBJZOPAV-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2OCCOC2=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)