CAS RN 25999-20-6
CAS RN 25999-20-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lasalocid A Sodium Salt Analytical Standard, MilliporeSigma™ Supelco™
Lasalocid is an antibacterial agent, belonging to the carboxylic polyether group of antibiotics, produced by the fermentation of Streptomyces lasaliensis. It is commercially available as a sodium salt and is reported to be used as a coccidiostat in poultry.