CAS RN 25913-05-7
CAS RN 25913-05-7
IUPAC Name:
4-(4-fluorobenzoyl)phenol
Synonyms:
4-(4-fluorobenzoyl)phenol
Molecular Weight (g/mol):
216.21
Molecular Formula:
C13H9FO2
InChi Key:
HLRVUOFDBXRZBI-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
4-Fluoro-4'-hydroxybenzophenone 98.0+%, TCI America™
CAS: 25913-05-7 Molecular Formula: C13H9FO2 Molecular Weight (g/mol): 216.211 MDL Number: MFCD00011484 InChI Key: HLRVUOFDBXRZBI-UHFFFAOYSA-N PubChem CID: 117666 IUPAC Name: (4-fluorophenyl)-(4-hydroxyphenyl)methanone SMILES: C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)F)O