accessibility menu, dialog, popup

UserName

CH3H3CH3CH3CNONHOI

CAS RN 25713-24-0

IUPAC Name: [4-(2-iodoacetamido)-2,2,6,6-tetramethylpiperidin-1-yl]oxidanyl
Synonyms: 4-(2-iodoacetamido)-2,2,6,6-tetramethylpiperidin-1-yloxidanyl
Molecular Weight (g/mol): 339.2
Molecular Formula: C11H20IN2O2
InChi Key: UCTVRHAKQRFPEZ-UHFFFAOYSA-N
SMILES: CC1(C)CC(CC(C)(C)N1[O])NC(=O)CI
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results