accessibility menu, dialog, popup

UserName

H3CNH2H3CCH3OO

CAS RN 2566-34-9

IUPAC Name: 2-methyl-2-(methylazaniumyl)propanoate
Synonyms: 2-methyl-2-(methylammonio)propanoate
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: DLAMVQGYEVKIRE-UHFFFAOYSA-N
SMILES: C[NH2+]C(C)(C)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results