accessibility menu, dialog, popup

UserName

CAS RN 25569-77-1

IUPAC Name: 4-fluorobenzoyl 4-fluorobenzoate
Synonyms: 4-fluorobenzoyl 4-fluorobenzoate
Molecular Weight (g/mol): 262.21
Molecular Formula: C14H8F2O3
InChi Key: BBLXFRIGTQYGOT-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results