accessibility menu, dialog, popup

UserName

CH3NH2CH3H3CH2N

CAS RN 25513-64-8

IUPAC Name: 2,2,4-trimethylhexane-1,6-diamine
Synonyms: 2,2,4-trimethylhexane-1,6-diamine
Molecular Weight (g/mol): 158.29
Molecular Formula: C9H22N2
InChi Key: JCUZDQXWVYNXHD-UHFFFAOYNA-N
SMILES: CC(CCN)CC(C)(C)CN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results