CAS RN 25498-49-1
IUPAC Name:
2-({1-[(1-methoxypropan-2-yl)oxy]propan-2-yl}oxy)propan-1-ol
Synonyms:
dowanol tpm
Molecular Weight (g/mol):
206.28
Molecular Formula:
C10H22O4
InChi Key:
GVZNXUAPPLHUOM-UHFFFAOYNA-N
SMILES:
COCC(C)OCC(C)OC(C)CO
1
–
1
of
1
results
1
Tripropylene Glycol Monomethyl Ether (mixture of isomer) 93.0+%, TCI America™
CAS: 25498-49-1 Molecular Formula: C10H22O4 MDL Number: MFCD00059142