accessibility menu, dialog, popup

UserName

CAS RN 254109-20-1

IUPAC Name: 4-amino-2',7'-difluoro-3',6'-dihydroxy-5-(methylamino)-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 4-amino-2',7'-difluoro-3',6'-dihydroxy-5-(methylamino)spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 412.35
Molecular Formula: C21H14F2N2O5
InChi Key: DIJCILWNOLHJCG-UHFFFAOYSA-N
SMILES: CNC1=C(N)C2=C(C=C1)C1(OC2=O)C2=CC(F)=C(O)C=C2OC2=CC(O)=C(F)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results