CAS RN 253342-48-2
CAS RN 253342-48-2
IUPAC Name:
4,4,5,5-tetramethyl-2-(3-methylphenyl)-1,3,2-dioxaborolane
Synonyms:
4,4,5,5-tetramethyl-2-(3-methylphenyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol):
218.1
Molecular Formula:
C13H19BO2
InChi Key:
XDKYCZBGHPGKEP-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
4,4,5,5-Tetramethyl-2-(m-tolyl)-1,3,2-dioxaborolane 98.0+%, TCI America™
CAS: 253342-48-2 Molecular Formula: C13H19BO2 Molecular Weight (g/mol): 218.10 MDL Number: MFCD05864303 InChI Key: XDKYCZBGHPGKEP-UHFFFAOYSA-N Synonym: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene, m-Tolylboronic Acid Pinacol Ester PubChem CID: 2760598 IUPAC Name: 4,4,5,5-tetramethyl-2-(3-methylphenyl)-1,3,2-dioxaborolane SMILES: CC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1