CAS RN 25294-65-9
IUPAC Name:
3-(4-bromophenyl)prop-2-ynoic acid
Synonyms:
3-(4-bromophenyl)prop-2-ynoic acid
Molecular Weight (g/mol):
225.04
Molecular Formula:
C9H5BrO2
InChi Key:
GZFOMZGALYVQDX-UHFFFAOYSA-N
SMILES:
OC(=O)C#CC1=CC=C(Br)C=C1
1
–
1
of
1
results
1
3-(4-Bromophenyl)propiolic Acid 98.0+%, TCI America™
CAS: 25294-65-9 Molecular Formula: C9H5BrO2 Molecular Weight (g/mol): 225.04 MDL Number: MFCD03230769 InChI Key: GZFOMZGALYVQDX-UHFFFAOYSA-N Synonym: (4-Bromophenyl)propargylic Acid, 3-(4-Bromophenyl)-2-propynoic Acid PubChem CID: 825805 IUPAC Name: 3-(4-bromophenyl)prop-2-ynoic acid SMILES: OC(=O)C#CC1=CC=C(Br)C=C1